Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIRC2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8278R

Description
Electrogenic metabolite transporter.Specifications
DIRC2 | |
Polyclonal | |
Unconjugated | |
Dirc2 | |
Dirc2; Disrupted in renal cancer protein 2; disrupted in renal carcinoma 2; disrupted in renal carcinoma 2 (human); Disrupted in renal carcinoma protein 2; Disrupted in renal carcinoma protein 2 homolog; RCC4; renal cell carcinoma 4; Slc49a4; Solute carrier family 49 member 4 | |
Rabbit | |
Protein A | |
RUO | |
84925 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96SL1 | |
Dirc2 | |
KLH conjugated synthetic peptide derived from human DIRC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction