Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Black 22, Thermo Scientific™

Catalog No. ac199470500
Click to view available options
Quantity:
50 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6473-13-8
Molecular Formula C44H32N13NaO11S3-2
Molecular Weight (g/mol) 1037.99
InChI Key RZEULCCAVHCFHK-KBDUNXJJSA-K
Synonym C.I. 35435
PubChem CID 71774637
IUPAC Name sodium;(3Z)-6-[(2,4-diaminophenyl)diazenyl]-3-[[4-[4-[(2Z)-2-[7-[(2,4-diaminophenyl)diazenyl]-1-oxo-3-sulfonatonaphthalen-2-ylidene]hydrazinyl]-2-sulfonatoanilino]phenyl]hydrazinylidene]-4-oxonaphthalene-2-sulfonate
SMILES C1=CC(=CC=C1NC2=C(C=C(C=C2)NN=C3C(=CC4=C(C3=O)C=C(C=C4)N=NC5=C(C=C(C=C5)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-])NN=C6C(=CC7=C(C6=O)C=C(C=C7)N=NC8=C(C=C(C=C8)N)N)S(=O)(=O)[O-].[Na+]

Specifications

Infrared Spectrum Authentic
Formula Weight 1083.98
UV Spectrometry Complies
Color Black
Quantity 50 g
Physical Form Powder
Chemical Name or Material Direct Black 22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.