Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Violet 1, TCI America™
SDP

Catalog No. C055125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 2586-60-9
Molecular Formula C32H22N6Na2O8S2
Molecular Weight (g/mol) 728.66
MDL Number MFCD00046486
InChI Key OSMMFJNWEWZQTR-UHFFFAOYSA-L
Synonym Diamine Violet N, Dianil Violet H, Chlorazol Violet N, Erie Violet 3R, Pontamine Violet N
PubChem CID 126957491
IUPAC Name disodium 6-imino-5-(2-{4'-[2-(2-imino-8-oxo-5-sulfonato-2,8-dihydronaphthalen-1-yl)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-yl)-4-oxo-4,6-dihydronaphthalene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=C2C=CC(=N)C(NNC3=CC=C(C=C3)C3=CC=C(NNC4=C5C(=O)C=CC(=C5C=CC4=N)S([O-])(=O)=O)C=C3)=C2C(=O)C=C1

Specifications

Quantity 25 g
Formula Weight 728.66
Physical Form Crystalline Powder
Chemical Name or Material Direct Violet 1
RTECSNumber QK1420000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.