Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Diridavumab Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542027
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Diridavumab is a biosimilar that targets influenza A virus hemagglutinin HA2 subunit from various strains.
Specifications
Diridavumab | |
Recombinant Monoclonal | |
Unconjugated | |
CR6261; CR-6261 | |
Influenza A virus hemagglutinin HA2 subunit from various strains. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1.41 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction