Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIS3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DIS3 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DIS3 Polyclonal specifically detects DIS3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DIS3 | |
Polyclonal | |
Rabbit | |
Human | |
2810028N01Rik, DIS3 mitotic control homolog (S. cerevisiae), dis3p, DKFZp667L1817, EC 3.1.13, EC 3.1.13.-, EC 3.1.26.-, EXOSC11, exosome complex exonuclease RRP44, exosome component 11, FLJ10484, KIAA1008RP11-342J4.3, MGC33035, mitotic control protein dis3 homolog, Protein DIS3 homolog, Ribosomal RNA-processing protein 44, RRP44bA555G22.1 | |
DIS3 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
22894 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSSNLCSLKCDVDRLAFSCIWEMNHNAEILKTKFTKSVINSKASLTYAEAQLRIDSANMNDDITTSLRGLNKLAKILKKRRIEKGALTLSSPEV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title