Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534549
Description
A suggested positive control is human ovary tissue lysate. PA5-34549 can be used with blocking peptide PEP-1592.
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
Specifications
DIS3 | |
Polyclonal | |
Unconjugated | |
DIS3 | |
2810028N01Rik; bA555G22.1; DIS3; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; DIS3 mitotic control homolog (S. cerevisiae); dis3p; EXOSC11; exosome complex exonuclease RRP44; exosome component 11; KIAA1008; mitotic control protein dis3 homolog; protein DIS3 homolog; RGD1304646; Ribosomal RNA-processing protein 44; RP11-342J4.3; RRP44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22894, 306103, 72662 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9CSH3, Q9Y2L1 | |
DIS3 | |
An 18 amino acid peptide near the amino terminus of human DIS3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction