Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558723 Supplier Invitrogen™ Supplier No. PA558723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAYILFVRKN AIVVLIPKYG LEGTVFFEEK DKPNPQLIYD DEIPSLKIED TVFHVFDKVK VKIMLDSSNL QHQKIRMSLV EPQIPGISIP TDTSN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DIS3
Gene Accession No. Q9Y2L1
Gene Alias 2810028N01Rik; bA555G22.1; DIS3; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; DIS3 mitotic control homolog (S. cerevisiae); dis3p; EXOSC11; exosome complex exonuclease RRP44; exosome component 11; KIAA1008; mitotic control protein dis3 homolog; protein DIS3 homolog; RGD1304646; Ribosomal RNA-processing protein 44; RP11-342J4.3; RRP44
Gene Symbols DIS3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DIS3. Recombinant protein control fragment (Product #RP-97922).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.