Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIS3L Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297612IR
Description
DIS3L Polyclonal specifically detects DIS3L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DIS3L | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DIS3 mitotic control homolog (S. cerevisiae)-like, DIS3-like exonuclease 1, EC 3.1.13, FLJ38088, KIAA1955EC 3.1.13.-, MGC4562 | |
Produced in rabbits immunized with E. coli-derived Human DIS3L fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
115752 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction