Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIS3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559739 Supplier Invitrogen™ Supplier No. PA559739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRNLLKDARH DCILFANEFQ QCCYLPRERG ESMEKWQTRS IYNAAVWYYH HCQDRMPIVM VTEDEEAIQQ YGSETEGVFV ITFKNYLDNF WPDLKAAHE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

DIS3L is a probable exonuclease. It has four isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DIS3L
Gene Accession No. Q8TF46
Gene Alias AV340375; DIS3 like exosome 3'-5' exoribonuclease; DIS3 mitotic control homolog (S. cerevisiae)-like; DIS3 mitotic control homolog-like; Dis3l; DIS3L1; DIS3-like exonuclease 1; DIS3-like exosome 3'-5' exoribonuclease; KIAA1955; RGD1308959
Gene Symbols DIS3L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DIS3L. Recombinant protein control fragment (Product #RP-98155).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.