Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIS3L2 Monoclonal Antibody (6C7B2)
GREENER_CHOICE

Catalog No. PIMA531718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531718 Supplier Invitrogen™ Supplier No. MA531718
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31718 has been tested in indirect ELISA.

DIS3L2 encodes a protein that is similar in sequence to 3'/5' exonucleolytic subunits of the RNA exosome. The exosome is a large multimeric ribonucleotide complex responsible for degrading various RNA substrates. Several transcript variants, some protein-coding and some not, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3L2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6C7B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene DIS3L2
Gene Accession No. Q8IYB7
Gene Alias 4930429A22Rik; 8030493P09Rik; DIS3 like 3'-5' exoribonuclease 2; DIS3 mitotic control homolog (S. cerevisiae)-like 2; DIS3 mitotic control homolog-like 2; DIS3 mitotic control-like 2; Dis3l2; DIS3-like 3'-5' exoribonuclease 2; DIS3-like exonuclease 2; FAM6A; family with sequence similarity 6, member A; hDIS3L2; PRLMNS; RGD1560168
Gene Symbols DIS3L2
Host Species Mouse
Immunogen Purified recombinant fragment of human DIS3L2 (AA: 27-250) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129563
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.