Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIS3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531273 Supplier Invitrogen™ Supplier No. PA531273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (90%), Rat (91%), Pig (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DIS3L2 encodes a protein that is similar in sequence to 3'/5' exonucleolytic subunits of the RNA exosome. The exosome is a large multimeric ribonucleotide complex responsible for degrading various RNA substrates. Several transcript variants, some protein-coding and some not, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3L2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DIS3L2
Gene Accession No. Q8IYB7
Gene Alias 4930429A22Rik; 8030493P09Rik; DIS3 like 3'-5' exoribonuclease 2; DIS3 mitotic control homolog (S. cerevisiae)-like 2; DIS3 mitotic control homolog-like 2; DIS3 mitotic control-like 2; Dis3l2; DIS3-like 3'-5' exoribonuclease 2; DIS3-like exonuclease 2; FAM6A; family with sequence similarity 6, member A; hDIS3L2; PRLMNS; RGD1560168
Gene Symbols DIS3L2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DIS3L2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129563
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.