Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIS3L2 Rabbit anti-Human, Mouse, Polyclonal, ProteinTech
SDP

Catalog No. 502238789 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8789 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8789 Supplier Proteintech Group Inc Supplier No. 288161AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

DIS3L2 Polyclonal antibody specifically detects DIS3L2 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen DIS3L2
Applications Western Blot
Classification Polyclonal
Concentration 600 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DIS3L2
Gene Accession No. Q8CI75, Q8IYB7
Gene Alias 4930429A22Rik; 8030493P09Rik; DIS3 like 3'-5' exoribonuclease 2; DIS3 mitotic control homolog (S. cerevisiae)-like 2; DIS3 mitotic control homolog-like 2; DIS3 mitotic control-like 2; Dis3l2; DIS3-like 3'-5' exoribonuclease 2; DIS3-like exonuclease 2; FAM6A; family with sequence similarity 6, member A; hDIS3L2; PRLMNS; RGD1560168
Gene Symbols DIS3L2
Host Species Rabbit
Immunogen DIS3L2 Fusion Protein Ag30634
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129563, 208718
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.