Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DISC1 Antibody, Novus Biologicals™
SDP

Catalog No. p-5064341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040775T 0.025 mL
NB11040775 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11040775T Supplier Novus Biologicals Supplier No. NB11040775SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

DISC1 Polyclonal specifically detects DISC1 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen DISC1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 2.5-5ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 24560582)
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Alias C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9
Gene Symbols DISC1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 700-800) of the mouse DISC1 protein. [Swiss-Prot# Q811T9]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27185
Test Specificity Mouse and rat. Immunogen sequence has 42% homology to human.
Target Species Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.