Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DISC1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040773H
Description
DISC1 Polyclonal specifically detects DISC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
DISC1 | |
Polyclonal | |
Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HRP | |
PBS with No Preservative | |
DISC1 | |
A synthetic peptide made to an internal region (within residues 400-500) of the rat DISC1 protein. [Swiss-Prot# Q810H6] | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
27185 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction