Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DISC1, p Ser710 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323278
Description
DISC1 Polyclonal antibody specifically detects DISC1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
DISC1 | |
Polyclonal | |
Western Blot 1:1000 | |
C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9 | |
Phospho-DISC1 (Ser-710) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-710 in mouse DISC1. This peptide sequence is well conserved in rat DISC1 (Ser-703), but has five amino acid differences with the conserved site in human DISC1 (Ser-713). [UniProt# Q811T9] | |
0.1 mL | |
Alzheimers Research, Cognition and Behavior, Neurodegeneration, Neuroscience | |
27185 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction