Learn More
Invitrogen™ DISC1 Polyclonal Antibody, DyLight™ 650

Description
The theoretical molecular weight of DISC1 is ~90 kDa. Due to alternative splicing, DISC1 may also run at ~75 and 100 kDa. Suggested positive control: human brain sections (neurons and glia).
Specifications
Specifications
| Antigen | DISC1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.85 mg/mL |
| Conjugate | DyLight 650 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | DISC1 |
| Gene Accession No. | Q810H6, Q811T9, Q9NRI5 |
| Gene Alias | C1orf136; DISC1; disrupted in schizophrenia 1; disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein; KIAA0457; RP4-730B13.1; SCZD9 |
| Gene Symbols | DISC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.