Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DISC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, liquid, OriGene

Rabbit Polyclonal Antibody
Supplier: OriGene TA890111

Description
DISC1 (Disrupted in Schizophrenia 1) is a multifunctional protein that interacts with multiple proteins of the centrosome and cytoskeletal system, and is involved in neuronal functions that are dependent upon proper cytoskeletal regulation, including neuronal migration, neurite architecture and intracellular transport. Regions of the brain which express DISC1 include the hippocampus, lateral septum, amygdala, cerebral cortex, cerebellum and paraventricular hypothalamus, and loss of DISC1 function results in differences in structural organization.The DISC1 gene locus is associated with patients afflicted with schizophrenia as a result of chromosomal translocations. DISC-1 encodes a large protein predicted to contain a globular N-terminal domain and a helical C-terminal domain, both of which have the potential to form interactions with other proteins. DISC-1 interacts with proteins involved in the centrosome and cytoskeletal system, including MIPT3, MAP1A and NUDEL; proteins which localize receptors to membranes, including alpha-actinin 2, beta 4-spectrin; and proteins which transduce signals from membrane receptors, including ATF-4 and ATF-5. Therefore, DISC-1 is thought to be involved in intracellular transport, neurite architecture and/or neuronal migration, all of which are thought to be pathogenic in the schizophrenic brain. Decreased expression of DISC1 in neurons caused an accelerated rate of neuronal integration, resulting in aberrant morphological development, suggesting that DISC1 plays a role in dendritic development and synapse formation. DISC-1 is thought to be involved in intracellular transport, neurite architecture and/or neuronal migration, all of which are thought to be pathogenic in the schizophrenic brain.Specifications
DISC1 | |
Polyclonal | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q810H6, Q811T9, Q9NRI5 | |
DISC1 | |
Recombit protein of human DISC1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DISC1 | |
C1orf136, SCZD9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244667, 27185, 307940 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction