Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C8 HPLC Column, 5 μm Particle Size

Catalog No. 111007440 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
25 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-7440 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-7440 Supplier MilliporeSigma™ Supelco™ Supplier No. 568323U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Discovery™ BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery™ BIO Wide Pore C5 and the Discovery™ BIO Wide Pore C18.

Discovery™ BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery™ BIO Wide Pore C5 and the Discovery™ BIO Wide Pore C18. The difference in hydrophobicity gives it unique selectivity relative to these other phases. It is ideal for peptide mapping because it provides complementary information compared to a C18 separation. Because of its intermediate hydrophobicity, we also recommend it for method development or scouting work. As with all Discovery™ BIO Wide Pore phases, the C8 phase gives efficient, symmetrical peaks, exceptional stability, long column lifetime, and LC-MS compatibility.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 25 cm
Particle Size 5 μm
Pore Size 300 Å
Surface Area 100 m2/g
Stationary Phase C8 (Octyl)
Product Line Discovery
Matrix C8 (Octyl) Phase/silica particle platform
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique Wide Pore HPLC
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.