Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size

Catalog No. 111020843 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
5 cm
10 cm
12.5 cm
15 cm
25 cm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
10 mm
21.2 mm
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-0843 5 cm 4.6 mm
11-100-8465 5 cm 2.1 mm
11-100-7417 10 cm 2.1 mm
11-100-7306 12.5 cm 2.1 mm
11-100-7292 15 cm 2.1 mm
11-100-7204 15 cm 3 mm
11-100-7013 25 cm 3 mm
11-100-7418 10 cm 4 mm
11-100-7307 12.5 cm 4 mm
11-100-7205 15 cm 4 mm
11-100-7014 25 cm 4 mm
11-100-7419 10 cm 4.6 mm
11-101-6245 12.5 cm 4.6 mm
11-100-7012 25 cm 4.6 mm
11-101-6199 25 cm 10 mm
11-100-5710 25 cm 21.2 mm
11-100-8673 2 cm 2.1 mm
11-100-7251 25 cm 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 11-102-0843 Supplier MilliporeSigma™ Supelco™ Supplier No. 504947
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use Discovery™ C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.

  • Excellent reproducibility
  • Exceptional peak shape for basic and acidic analytes
  • Stable, low-bleed LC-MS separations
  • Separation of peptides and small proteins
  • Lower hydrophobicity than many comparable C18 columns, providing faster analysis
TRUSTED_SUSTAINABILITY

Specifications

Column Format HPLC Column
Diameter (Metric) 4.6 mm
Length (Metric) 5 cm
Max. Pressure 400 bar (5801 psi)
Particle Size 5 μm
pH 2 to 8
Pore Size 180 Å
Surface Area 200 m2/g
Temperature ≤70°C
USP Type USP L1
Stationary Phase C18 (Octadecyl)
Product Line Discovery
Matrix C18 (Octadecyl) Phase/Silica Gel, High Purity, Spherical Particle Platform
Packing Material Silica gel, high purity, spherical particle platform
Carbon Load 12%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique HPLC
Product Type HPLC Column
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.