Learn More
MilliporeSigma™ Supelco™ Discovery™ DPA-6S SPE Tube , Bed Wt. 250 mg
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
$210.00 - $241.00
Specifications
Column Phase DPA-6 S
Column Type SPE
Description 4.5 - 7.5 (surface pH)
Bed Weight 250 mg
For Use With (Application) Food and Beverages
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity & Availability  
11-100-4354
View Documents
MilliporeSigma™ Supelco™
52625U
3 mL 54/Pk.
Each for $241.00
Only null left
Add to Cart
11-100-4462
View Documents
MilliporeSigma™ Supelco™
52626U
6 mL 30/Pk.
Each for $210.00
Only null left
Add to Cart
Description
Description
Retention Mechanism: Reversed-phaseSample Matrix Compatibility: Aqueous or methanolic solutions. Polyamide Resin: Particle Size: 50-160 μm, Surf pH: 4.5-7.5, Density: 0.2-0.3 cm3/g, Water Content: < 5 %. Used to adsorb polar compounds (-OH groups, esp. phenolic compounds) from aqueous or methanolic solutions under the reversed-phase mechanism through strong hydrogen bonding between compound hydroxyl groups and amide groups of the resin. Useful for extracting tannins, chlorophyll, humic acid, pharmacologically active terpenoids, flavanoids, gallic acid, catechol A, protocatechuic acid, and phloroglucinol. Also useful for extracting aromatic carboxylic acids and nitroaromatic compounds. Irreversibly retains quinones
Specifications
Specifications
DPA-6 S
4.5 - 7.5 (surface pH)
Food and Beverages
Solid Phase Extraction Column
SPE
250 mg
Discovery
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.