Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ DSC-Si SPE Tube, Bed Wt. 500 mg
Shop All MilliporeSigma Supelco Products
Click to view available options
Volume (Metric):
3 mL
6 mL
Quantity:
30/Pk.
54/Pk.
Description
Retention Mechanism: Normal phaseSample Matrix Compatibility: Organic solvents, oils, and lipids. Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques. Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions. The most polar normal phase sorbent available. Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents
Specifications
Specifications
| Column Phase | DSC-Si |
| Column Type | SPE |
| Bed Weight | 500 mg |
| For Use With (Application) | Food and Beverages |
| Product Line | Discovery |
| Type | Solid Phase Extraction Column |
| Volume (Metric) | 3 mL |
| Material | PE Frit |
| Pore Size | 70 Å |
| Quantity | 54/Pk. |
| Show More |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction