Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Great Inventions with Bill Nye: Scientifc Advances in Our Homes
SCI_ED

Catalog No. s95451dvd
Click to view available options
Grade:
6 to 12

See how appliances and other conveniences at home have grown into the familiar forms of today

Bill Nye explains the technology of the house and home, from plumbing and paints to microwave ovens and frozen foods.

  • Grades 6-12
  • 56 mins.

Specifications

Grade 6 to 12
Run Time 56 min.
Includes Marc Record, Teacher's Guide
Format DVD
Copyright Year 2007
Name Greatest Inventions with Bill Nye: Scientific Advances in Our Homes DVD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.