Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Physical Science: The Basics, Grade 6 to 8
SCI_ED

Catalog No. s90451dvd
Click to view available options
Grade:
6 to 8

From electricity and magnetism to light and sound

Introduce young students to a wide variety of physical science topics with this award-winning series.

The title includes six scientific literacy segments to aid classroom teachers along with the following 12 student video segments:

  • Light and Heat
  • Friction
  • Calculating Average Speed
  • Roller Coasters
  • The Eye
  • Heat
  • Electricity
  • Magnetism
  • Chemical Properties of Mixtures
  • Polymers
  • Energy Transfer
  • Constant Speed

Order Info

Grade 6 to 6. Running time: 3 hours.

Specifications

Grade 6 to 8
Copyright Year 2005
Run Time 180 min.
Format DVD
Name Physical Science: The Basics 6-8 DVD
Includes Marc Record, Teacher's Guide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.