Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Understanding Robots DVD
SCI_ED

Catalog No. s14185
Click to view available options
Grade:
6 to 12

Alice Bot-Named most human computer on Earth

Specifications

Grade 6 to 12
Copyright Year 2006
Format DVD
Run Time 30 min.
Name Understanding Robots
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.