Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dishevelled-2 Antibody (OTI2D11), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270611AF350
Description
Dishevelled-2 Monoclonal specifically detects Dishevelled-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Dishevelled-2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
DVL2 | |
Recombinant protein fragment corresponding to amino acids 1-320 of human Dishevelled-2 (NP_004413) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI2D11 | |
Immunohistochemistry | |
dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2 | |
Mouse | |
0.1 mL | |
1856 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction