Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DISP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518257
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518257 Supplier Invitrogen™ Supplier No. PA518257
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP1
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DISP1
Gene Accession No. Q96F81
Gene Alias 1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866
Gene Symbols DISP1
Host Species Goat
Immunogen Synthetic peptide sequence (SELSGESLLIKTL) corresponding to the C-terminus amino acids of DISP1 (aa 1512-1524).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.