Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DISP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572744
Description
DISP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo.
Specifications
DISP1 | |
Polyclonal | |
Unconjugated | |
DISP1 | |
1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68897, 84976 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3TDN0, Q96F81 | |
DISP1 | |
DISP1 antibody for a 16 amino acid synthetic peptide near the amino terminus of human DISP1. The immunogen is within amino acids 20 - 70 of DISP1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction