Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DISP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584065 Supplier Invitrogen™ Supplier No. PA584065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKTNVHSLQR SIEEHLPKMA EPSSFVCRST GSLLKTCCDP ENKQRELCKN RDVSNLESSG GTENKAGGKV ELSLSQTDAS VNSE Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 40%.

The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DISP1
Gene Accession No. Q96F81
Gene Alias 1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866
Gene Symbols DISP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DISP1. Recombinant protein control fragment (Product #RP-99799).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84976
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.