Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DISP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563790 Supplier Invitrogen™ Supplier No. PA563790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIGSKLVVWR ALQALTGPRK LLFLSPDLEL NSSSSHNTLR PAPRGSAQES AVRPRRMVEP LEDRRQENFF CGPPEKSYAK LVFMSTSSGS LWNLHAIHSM CRMEQDQIR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.

DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DISP2
Gene Accession No. A7MBM2
Gene Alias AI840597; B230210L08Rik; C15orf36; DISP2; dispatched B; dispatched homolog 2; dispatched homolog 2 (Drosophila); dispatched RND transporter family member 2; DISPB; HsT16908; KIAA1742; LINC00594; mKIAA1742; Protein dispatched homolog 2
Gene Symbols DISP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DISP2. Recombinant protein control fragment (Product #RP-98095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.