Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DISP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981495
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1495 Supplier Bioss Supplier No. BS11588R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Disp2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the Disp proteins have been implicated in these pathways. Recently, it has been shown that Disp2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased Disp2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by Disp2 mutants. At least two isoforms of Disp2 are known to exist. This Disp2 antibody is predicted to not cross-react with Disp1 or Disp3.

Specifications

Antigen DISP2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DISP2
Gene Accession No. A7MBM2
Gene Alias AI840597; B230210L08Rik; C15orf36; DISP2; dispatched B; dispatched homolog 2; dispatched homolog 2 (Drosophila); dispatched RND transporter family member 2; DISPB; HsT16908; KIAA1742; LINC00594; mKIAA1742; Protein dispatched homolog 2
Gene Symbols DISP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DISP2 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85455
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.