Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MediPurpose™ SurgiLance™ Lite Safety Lancets
SDP

Catalog No. 22000233
Click to view available options
Depth (English) Needle:
1.4mm
1.8mm
2.2mm
Color:
Gray
Orange
Yellow
Flow Type:
Low (2 to 5μL)
Low (5 to 10μL)
Medium (10 to 20μL)
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for users with small hands.

  • Eco-friendly design
  • Single use design
  • 28-gauge needle
  • Easy to use
  • Minimizes pain

Specifications

Disposable Single-use
Model SurgiLance™ Lite
Needle Gauge 28G
For Use With (Application) Glucose test
Product Type Safety Lancet
Color Yellow
Depth (English) Needle 1.4mm
Flow Type Low (2 to 5μL)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.