Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse Blue 14, MP Biomedicals™

Catalog No. icn15787625 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 g

Catalog No. ICN15787625

Supplier: MP Biomedicals, Inc 0215787625

Chemical Identifiers

CAS 2475-44-7
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.30
MDL Number MFCD00001198
InChI Key QOSTVEDABRQTSU-UHFFFAOYSA-N
Synonym C.I. 61550
PubChem CID 17189
IUPAC Name 1,4-bis(methylamino)-9,10-dihydroanthracene-9,10-dione
SMILES CNC1=CC=C(NC)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Formula Weight 266.3
Quantity 25 g
Physical Form Powder
Chemical Name or Material Disperse Blue 14
Recommended Storage Store at room temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.