Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VELP Scientifica™ Dispersing Tool
SDP

Dispersing tool, suitable for emulsions and suspensions that require high final fineness.

Supplier:  VELP Scientifica™ A00000462

 View more versions of this product

Catalog No. 08-418-526


Only null left
Add to Cart

Description

Description

SAMPLES AND APPLICATIONS

  • Preparation of foamy mixtures (ice cream, butter, margarine, icing) and spreadable creams
  • Preparation of tablets coatings
  • Preparation of pharmaceutical ointments and creams
  • Preparation of cosmetic products (lip balm, soap, shampoo, conditioner, creams, lipstick, mascara)
  • Preparation of pigments, inks, paints
  • Preparation of biodiesel and dispersion of petroleum derivatives (e.g., asphalt)
Specifications

Specifications

Dispersing Tool
40 to 175 mm
Stainless Steel
Fibrous Sample Disperser
50 to 2000 mL
Autoclavable
Stainless Steel
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.