Description
- The filter’s 30 mm ID opening allows for an easy, air-tight fit to the flow head.
- The filters are single-person use only. We recommend one filter for every student who will be using the spirometer.
Specifications
Specifications
| Description | Disposable Bacterial Filter |
| Quantity | 30/Pk. |
| For Use With (Equipment) | Spirometer |