Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Disposable Base Molds
Shop All Epredia Products
Click to view available options
Dimensions (L x W):
15 x 15 mm
24 x 24 mm
30 x 24 mm
37 x 24 mm
Description
- Cleaning associated with metal molds is not required
- Transparency facilitates tissue orientation
Ordering Information:
- Sold in cases of four boxes.
Specifications
Specifications
Product Type | Base Mold |
Material | Plastic |
Dimensions (L x W) | 24 x 24 mm |
Disposable | Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction