Description
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
- Shell offers built-in chin protection and extended top-of-head coverage
- Adjustable headgear tilts visor into 2784 possible positions
- Ratcheting headgear features a locking mechanism for secure fit
- Comfortable cell foam on back of headgear with breathable, removable, washable headband
- 100% dielectric; no metal parts
Replacement Visors
- Clear lens suits most indoor work applications
- Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
Specifications
| Lens Coating | Uncoated |
| Certifications/Compliance | ANSI Z87.1-2010, CSA Z94.3 |
| Lens Tint | Clear |
| Product Type | Face Shield |
| Frame Color | Black |
| Lens Material | Polycarbonate |