Learn More
Fisherbrand™ Disposable Inoculating Loops
Shop All Fisherbrand Products
Click to view available options
Volume:
1 μL
10 μL
Sterility:
Non-sterile
Sterile
Quantity:
Cs of 1000 (40 Pks. of 25)
Cs of 1000 (50 Pks. of 20)
Cs of 125 (5 Pks. of 25/ Individually Packed)
Cs of 250 (5 Pks. of 50/ Individually Packed)
Description
- Ideal for anaerobic microbiology applications where deep agar inoculation is required
- Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
- High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
- Rigid enough to penetrate the media for subsurface inoculation
- Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
- Can be used under hoods without danger
- Color-coded for ease of size identification
Specifications
Specifications
Disposable | Single-use |
Volume | 1 μL |
Material | Polystyrene |
Sterility | Non-sterile |
Color | White |
Quantity | Cs of 1000 (50 Pks. of 20) |
Autoclavable | Not Autoclavable |