Learn More
GVS Disposable Inoculating Loop and Needle
Catalog No. 09316824 Shop All Osmonics Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Color:
Blue
Neutral
Material:
ABS
AS
Packaging Type:
1/Peel Pack
10/Zip-Lock Pack
20/Zip-Lock Pack
5/Zip-Lock Pack
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Color Material Packaging Type
09-316-824 1 μL Neutral AS 1/Peel Pack
09-316-796 10 μL Blue ABS 1/Peel Pack
09-316-798 10 μL Blue AS 5/Zip-Lock Pack
09-317-085 1 μL Neutral ABS 1/Peel Pack
09-317-086 1 μL Neutral AS 5/Zip-Lock Pack
09-317-084 10 μL Blue AS 10/Zip-Lock Pack
09-317-340 1 μL Neutral AS 20/Zip-Lock Pack
09-317-342 10 μL Blue ABS 20/Zip-Lock Pack
09-316-756 10 μL Blue AS 1/Peel Pack
09-316-755 1 μL Neutral ABS 10/Zip-Lock Pack
09-316-757 1 μL Neutral AS 10/Zip-Lock Pack
09-316-999 10 μL Blue ABS 5/Zip-Lock Pack
09-316-992 1 μL Neutral ABS 20/Zip-Lock Pack
09-317-174 1 μL Neutral ABS 5/Zip-Lock Pack
09-317-171 10 μL Blue ABS 10/Zip-Lock Pack
09-317-180 10 μL Blue AS 20/Zip-Lock Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 09-316-824 Supplier GVS Supplier No. CELCUCG2121X0044S
Only null left
Add to Cart
Add to Cart
GVS Disposable Inoculating Loops and Needles available in 1 and 10 μL with different packing for choices.
  • Smooth Surface – Ensures even and consistent streaking
  • Color-Coded Sizes – Allows quick and easy identification
  • Ergonomic Polygonal Shaft – Provides a secure grip for easy operation and precise directional control
  • Loop Options – Available in both rigid and flexible types upon request
  • Secure Packaging – Supplied in individual peel packs (easy peel-open) and zip-lock packs (tamperproof and re-sealable) to minimize the risk of contamination
Specifications
Volume 1 μL
Material AS
Sterility Sterile
Size Standard
Color Neutral
Product Type Inoculating Loop and Needle
Packaging Type 1/Peel Pack