Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Disposable Manual 25mL Reagent Reservoirs
GSA_VA

Catalog No. p-7078489
Click to view available options
Packaging:
100 reservoirs/Pk.
5 reservoirs per bag
Pk of 20
No. per Pack:
10 bags/Pk.
20
Bulk
Sterility:
Non-sterile
Sterile
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed with angled sides and V-shaped bottom.

  • Made from polystyrene
  • With graduation marks

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque-white
Capacity (Metric) 25 mL
Material Polystyrene
Sterility Sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Height (Metric) 24.13 mm
Length (Metric) 132.33 mm
No. per Pack 10 bags/Pk.
Packaging 5 reservoirs per bag
Quantity 50/Pk.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.