Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tians Disposable Polypropylene Coveralls
Shop All Tians International Products
Click to view available options
Size:
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Economical white polypropylene coverall is ideal low-cost alternative for general protection in nonhazardous environments
- Cool, comfortable, and lightweight
- Zipper front, elastic wrist and ankles, and folded collar
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 5X-Large |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction