Learn More
StatLab™ Dissect Aid™
GSA_VASDP
Catalog No. p-7227183
Click to view available options
Quantity:
2.5 gal.
5 gal.
4 x 1 gal.
Packaging Type:
Bottle
Cube
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
A fixative which helps make lymph nodes more identifiable during pathology examination.
  • Turns lymph nodes pale to white so they stand out from other tissue, while firming the nodes for touch identification when performing gross examination
  • Dramatically improves lymph node recovery or harvesting
  • Ideal fixative for fatty tissues
  • Formulation has helped in diagnosis for the past 30 years
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Packaging Type Cube
Quantity 2.5 gal.
Formulation Ethyl Alcohol, Acetic Acid, Methyl Alcohol, Formaldehyde, Isopropyl Alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.