Learn More
Mopec™ Dissecting Board with 12-inch Legs
SDP
Catalog No. 22444329 Shop All Mopec Products
Change view
Click to view available options
Color:
Dark Blue
Light Blue
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-444-329 Light Blue
22-444-326 White
22-444-338 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-444-329 Supplier Mopec™ Supplier No. BC060
Add to Cart
Add to Cart
Mopec Dissecting Board with 12-inch Legs is ideal for pathology specimen dissection
  • Easy to clean
  • Safe, rounded edges
  • Ergonomic, multi-user work surface
  • Optional removable legs
  • Non-skid feet
Specifications
Product Type Dissecting Board
Color Light Blue
Description 12-inch legs
Material Polyethylene
Dimensions (L x W) 23 x 16 in.