Description
- Easy to clean
- Safe, rounded edges
- Ergonomic, multi-user work surface
Specifications
Specifications
| Color | Dark Blue |
| Description | Dissecting Board |
| Material | Polyethylene |
| Dimensions (L x W) | 23 x 16 in. |