Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Dissecting Pan Wax
SDPSCI_ED

Catalog No. S89560 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S89560 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89560 Supplier United Scientific™ Supplier No. DSPA05WAX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

United Scientific™ Dissecting Pan Wax contains five pounds of black wax for dissecting trays.

Wax for dissecting trays

  • 5 lb. of Wax
  • Color: Black
  • For use with most types of dissecting pans

Specifications

Product Type Dissecting Wax
Autoclavable Not Autoclavable
Color Black
Material Wax
Weight (English) 5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.