Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Dissolved Oxygen Test Kit
SDPSCI_ED

Catalog No. S94979 Shop All LaMotte Company Products
Click to view available options
No. of Tests:
50

Includes

Five reagents; makes 50 tests at 10 ppm

Learn how to conduct oxygen-level tests for plants and animals with the Dissolved Oxygen Kit.

  • Direct-reading titrator test measures levels of oxygen.
  • Indicates whether a body of water can sustain healthy aquatic plants and animals.
  • Low oxygen levels signal contamination from organic wastes.
  • Measures 0 to 10 ppm in 0.2 ppm increments.
  • Azide modification of Winkler method.
  • Kit Includes: 30 mL Manganous Sulfate Solution, 30 mL Alkaline Potassium Iodide Azide, 30 mL Sulfuric Acid, 60 mL 0.025 N Sodium Thiosulfate, 30 mL Starch Indicator Solution, Direct Reading Titrator, 5-10-12.9-15-20-25 mL Glass Test Tube with Cap, 60 mL Water Sampling Glass Bottle

Specifications

Volume (Metric) 200 mL
For Use With (Application) Water Testing
For Use With (Equipment) Dissolved Oxygen Kit
Includes 5 Reagents, Direct Reading Titrator, Glass Test Tube with Cap, Water Sampling Glass Bottle
No. of Tests 50
Type Dissolved Oxygen Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.