Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Acids and Bases
GREENER_CHOICESCI_ED

Catalog No. S08191 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08191 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08191 Supplier Innovating Science™ Supplier No. IS1500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An acid and base introduction unit, litmus paper tests

  • Objectives:
    • Acid and base introduction
    • Test a dilute acid and dilute base with neutral litmus paper
    • Learn how pH strips indicate if something is an acid or base
    • Test common household items and determine if they are acids or bases
  • Includes:
    • 10 mL 0.01M HCl
    • 10 mL 0.01M NaOH
    • 10 mL distilled water
    • 10 mL vinegar
    • 10 mL household ammonia
    • 10 mL liquid hand soap
    • 10 mL lemon juice
    • 1 Pk. neutral litmus paper
    • 2 spot plates
  • Notes:
    • Extra litmus paper included if students want to test more items
    • Contains an instruction manual and enough materials for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.