Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Acids, Bases and the pH Scale
GREENER_CHOICESCI_ED

Catalog No. S08194 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08194 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08194 Supplier Innovating Science™ Supplier No. IS1503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Discusses differences between properties of acids and bases, the role of hydrogen and hydroxide ions in acidic and basic substances, and the pH scale with regards to acid and base strength

  • Objectives:
    • Examine effects of acids and bases on several pH indicators
    • Determine pH of several common household materials
    • Use knowledge gained to determine the identity of four unknown solutions
  • Includes:
    • 5 mL deionized water
    • 5 mL dilute acid (0.1 M HCl)
    • 5 mL dilute base (0.1 M NaOH)
    • 5 mL litmus blue (0.5%)
    • 5 mL methyl red (0.02%)
    • 5 mL bromothymol blue (0.5%)
    • 5 mL phenolphthalein (1%)
    • 5 mL vinegar
    • 5 mL household ammonia
    • 5 mL soap solution
    • 5 mL filtered water
    • 5 mL vitamin C solution
    • 5 mL unknown solution No. 1 (water)
    • 5 mL unknown solution No. 2 (dilute base)
    • 5 mL unknown solution No. 3 (phenolphthalein)
    • 5 mL unknown solution No. 4 (dilute acid)
    • 1 wide-range pH test strips (50 Pk.)
    • 2 reaction trays
  • Note:
    • Instruction manual and enough materials for two groups included

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.