Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Forensic Chemistry - Drug Detection
GREENER_CHOICESCI_ED

Catalog No. S08200 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08200 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08200 Supplier Innovating Science™ Supplier No. IS1509
Add to Cart
Add to Cart

Students work as forensic toxicologists to perform chemical tests in order to determine if all ingredients are what they should be

  • Objectives:
    • Work as a forensic toxicologist to determine if someone tainted the cafeteria's chili
    • Determine if any ingredients were substituted for aspirin (stolen form the nurse's office)
    • Perform chemical tests on cafeteria ingredients and control aspirin
  • Includes:
    • 4 g acetylsalicylic acid (control)
    • 4 g corn starch
    • 4 g salt
    • 4 g sugar
    • 4 mL sodium hydroxide, 1M
    • 4 mL ferric nitrate, 0.2M
    • 4 mL Lugol's iodine
    • 4 mL deionized water
    • 1 Pk. toothpicks
    • 4 plastic scoops
    • 2 microreaction plates
  • Notes:
    • Contains an instruction manual and enough material for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Forensic Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.