Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Kidneys And Blood Filtration
GREENER_CHOICESCI_ED

Catalog No. S08197 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08197 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08197 Supplier Innovating Science™ Supplier No. IS1506
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Understand kidneys and their role in blood filtration, determine if blood filtration has occurred based on a simulation

  • Objectives:
    • Understand the role of the kidney in blood filtration and waste removal
    • Understand the function of nephrons and their structure
    • Create an artificial kidney model to filter a simulated blood solution
    • Determine if filtration of simulated blood occurred
    • Perform chemical tests on filtrate to detect waste material that may have been removed by artificial kidney
  • Includes:
    • 10 mL simulated unfiltered blood
    • 250 mL deionized water
    • 2 pieces dialysis tubing
    • 2 plastic cups
    • 2 pipettes
    • 4 salt test strips
    • 4 urea test strips
  • Notes:
    • Contains instruction manual and enough material for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Kidney and Blood Filtration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.