Learn More
Ritmed™ DisTech™ Isolation Gown
Catalog No. 19910742 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium/Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-742 X-Large
29-636-8443 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-742 Supplier Ritmed™ Supplier No. D8010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolation gowns feature thumb loop cuffs to keep sleeves in place. Not ISO-rated.
  • Designed to protect healthcare professionals and patients against cross-contamination
  • Features comfortable fit with thumb loop cuffs
  • Sewn seams provide for superior strength
  • Available in assorted sizes
  • Single-use disposable
  • Made of spunbond polypropylene (latex-free)
Specifications
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Size X-Large
Color Yellow
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Model D-8010
Sleeve Style Long Sleeve
Includes 50 Gowns/Cs.
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov